siRNA Details
| Gene Symbol: Klhdc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033039.2 | NP_001028211.1 | 1, 2 | 661 | ||
| GenBank mRNA | AK076812.1 | - | 1, 2 | 384 | ||
| AK140290.1 | - | 1, 2 | 662 | |||
| BC141196.1 | - | 1, 2 | 574 | 9055808 | ||

Klhdc9
Genomic Map