siRNA Details
| Gene Symbol: 1190003K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195435.1 | NP_001182364.1 | 2 | 437 | ||
| GenBank mRNA | AK004471.1 | - | 2 | 438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1190003K10Rik| Gene Symbol: 1190003K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195435.1 | NP_001182364.1 | 2 | 437 | ||
| GenBank mRNA | AK004471.1 | - | 2 | 438 | ||