siRNA Details
| Gene Symbol: Hint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026871.1 | NP_081147.1 | 5 | 458 | ||
| GenBank mRNA | AF356874.1 | - | 5 | 463 | ||
| AK004497.1 | - | 5 | 460 | |||
| AK170135.1 | - | 4 | 1774 | |||
| BC086940.1 | - | 5 | 458 | 6775318 | ||

Hint2
Genomic Map