siRNA Details
| Gene Symbol: 1190005I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197988.1 | NP_932105.2 | 4 | 677 | ||
| GenBank mRNA | AK004494.1 | - | 4 | 626 | ||
| BC027666.1 | - | 4 | 677 | 1245153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1190005I06Rik| Gene Symbol: 1190005I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197988.1 | NP_932105.2 | 4 | 677 | ||
| GenBank mRNA | AK004494.1 | - | 4 | 626 | ||
| BC027666.1 | - | 4 | 677 | 1245153 | ||