siRNA Details
| Gene Symbol: Ptcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026873.2 | NP_081149.1 | 2 | 130 | ||
| GenBank mRNA | AK004502.1 | - | 2 | 140 | ||
| AK144396.1 | - | 2 | 142 | |||
| BC016563.1 | - | 2 | 132 | |||
| BC025110.1 | - | 2 | 130 | 3987809 | ||

Ptcd2
Genomic Map