siRNA Details
| Gene Symbol: Aspscr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026877.2 | NP_081153.1 | 11 | 1373 | ||
| NM_198223.2 | NP_937866.1 | 10 | 1152 | |||
| GenBank mRNA | AK004509.1 | - | 5 | 385 | ||
| AK088197.1 | - | 11 | 1211 | |||
| AY349132.1 | - | 9 | 1142 | |||
| AY349133.1 | - | 11 | 1290 | |||
| BC019177.1 | - | 11 | 1316 | 5008127 | ||
| BC022115.1 | - | 11 | 1320 | 5101238 | ||
| BC031153.1 | - | 11 | 1305 | 4976569 | ||

Aspscr1
Genomic Map