siRNA Details
| Gene Symbol: Tmco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039483.1 | NP_001034572.1 | 5 | 498 | ||
| GenBank mRNA | AK004504.1 | - | 5 | 340 | ||
| AK015068.1 | - | 5 | 789 | |||
| AK037709.1 | - | 5 | 433 | |||
| AK178225.1 | - | 2 | 102 | |||
| BC011457.1 | - | 5 | 372 | 3585288 | ||
| BC019761.1 | - | 5 | 416 | 5009960 | ||
| BC020098.1 | - | 5 | 428 | 4019484 | ||

Tmco1
Genomic Map