siRNA Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026883.3 | NP_081159.1 | 6 | 612 | ||
| GenBank mRNA | AK005211.1 | - | 6 | 1004 | ||
| AK182306.1 | - | 2 | 140 | |||
| BC019498.1 | - | 6 | 970 | 4460879 | ||
| BC057658.1 | - | 6 | 651 | 3153717 | ||
| BC061033.1 | - | 6 | 612 | 6590939 | ||

Fam216a
Genomic Map