siRNA Details
| Gene Symbol: Fam57b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146347.1 | NP_001139819.1 | 3 | 697 | ||
| NM_026884.1 | NP_081160.1 | 7 | 810 | |||
| NM_029978.1 | NP_084254.1 | 4 | 1249 | |||
| GenBank mRNA | AK005271.1 | - | 4 | 561 | ||
| AK131642.1 | - | 4 | 1248 | |||
| BC055444.1 | - | 3 | 698 | 4502433 | ||
| BC093505.1 | - | 835 | 4503230 | |||

Fam57b
Genomic Map