siRNA Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026886.3 | NP_081162.1 | 9 | 1147 | ||
| GenBank mRNA | AK053836.1 | - | 9 | 879 | ||
| AK129457.1 | - | 1 | 229 | |||
| AK165719.1 | - | 10 | 881 | |||
| BC056392.1 | - | 9 | 1137 | 6855144 | ||
| JN860221.1 | - | 9 | 794 | |||

Srrm4
Genomic Map