siRNA Details
| Gene Symbol: Srrm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026886.3 | NP_081162.1 | 11, 12 | 1743 | ||
| GenBank mRNA | AK053836.1 | - | 11, 12 | 1475 | ||
| AK129457.1 | - | 3, 4 | 825 | |||
| BC056392.1 | - | 11, 12 | 1733 | 6855144 | ||
| JN860221.1 | - | 11, 12 | 1390 | |||

Srrm4
Genomic Map