siRNA Details
| Gene Symbol: SCGB2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002407.2 | NP_002398.1 | 2 | 176 | ||
| GenBank mRNA | AF071219.1 | - | 2 | 176 | ||
| AJ224173.1 | - | 2 | 152 | |||
| BC062218.1 | - | 2 | 175 | 30351353 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCGB2A1| Gene Symbol: SCGB2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002407.2 | NP_002398.1 | 2 | 176 | ||
| GenBank mRNA | AF071219.1 | - | 2 | 176 | ||
| AJ224173.1 | - | 2 | 152 | |||
| BC062218.1 | - | 2 | 175 | 30351353 | ||