siRNA Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026896.5 | NP_081172.2 | 2 | 261 | ||
| NM_029615.1 | NP_083891.1 | 2 | 261 | |||
| GenBank mRNA | AK005251.1 | - | 2 | 236 | ||
| AK009764.1 | - | 2 | 242 | |||
| AK147181.1 | - | 2 | 258 | |||
| BC016537.1 | - | 2 | 252 | 3585027 | ||
| CJ072886.1 | - | 2 | 262 | |||

Med27
Genomic Map