siRNA Details
| Gene Symbol: Wdr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185162.1 | NP_001172091.1 | 2 | 451 | ||
| NM_026898.2 | NP_081174.1 | 2 | 683 | |||
| GenBank mRNA | AK005105.1 | - | 2 | 650 | ||
| BC028850.1 | - | 1 | 415 | 3987279 | ||

Wdr53
Genomic Map