siRNA Details
| Gene Symbol: Ssu72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026899.3 | NP_081175.2 | 2 | 731 | ||
| GenBank mRNA | AK004446.1 | - | 2 | 421 | ||
| AK005220.1 | - | 2 | 422 | |||
| AK019168.1 | - | 2 | 187 | |||
| AK148131.1 | - | 2 | 495 | |||
| AK148783.1 | - | 1 | 28 | |||
| AK184265.1 | - | 1 | 65 | |||
| BC016544.1 | - | 2 | 411 | 3497878 | ||

Ssu72
Genomic Map