siRNA Details
| Gene Symbol: Zfp580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026900.1 | NP_081176.1 | 2 | 557 | ||
| GenBank mRNA | AB221670.1 | - | 2 | 536 | ||
| AK005257.1 | - | 2 | 552 | |||
| BC066001.1 | - | 2 | 505 | 5700640 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp580| Gene Symbol: Zfp580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026900.1 | NP_081176.1 | 2 | 557 | ||
| GenBank mRNA | AB221670.1 | - | 2 | 536 | ||
| AK005257.1 | - | 2 | 552 | |||
| BC066001.1 | - | 2 | 505 | 5700640 | ||