siRNA Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026917.5 | NP_081193.2 | 6 | 938 | ||
| GenBank mRNA | AB088044.1 | - | 5 | 704 | ||
| AK007355.1 | - | 7 | 1410 | |||
| AK018755.1 | - | 6 | 905 | |||
| AK080541.1 | - | 6 | 904 | |||
| AK169197.1 | - | 885 | ||||
| AY166672.1 | - | 5 | 704 | |||
| BC005531.1 | - | 7 | 965 | |||
| BC018334.1 | - | 5 | 774 | |||
| BC025126.1 | - | 6 | 923 | 3669723 | ||
| BC052464.1 | - | 6 | 843 | 6834763 | ||

Zdhhc3
Genomic Map