siRNA Details
Gene Symbol: Tmem258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163431.1 | NP_001156903.1 | 3 | 237 | ||
NM_026919.1 | NP_081195.1 | 3 | 208 | |||
GenBank mRNA | AK007351.1 | - | 3 | 193 | ||
AK145833.1 | - | 3 | 201 | |||
AV456553.1 | - | 3 | 179 | |||
BC058335.1 | - | 3 | 164 | |||
BC085316.1 | - | 2 | 834 |
