siRNA Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022885.2 | NP_075023.2 | 14, 15 | 2233 | ||
| GenBank mRNA | AF233321.1 | - | 7, 8 | 1252 | ||
| AF461761.1 | - | 14, 15 | 2132 | |||
| AK051183.1 | - | 14, 15 | 2233 | |||
| AK088004.1 | - | 13, 14 | 1977 | |||
| AK090176.1 | - | 14, 15 | 2142 | |||
| AK166006.1 | - | 14, 15 | 2043 | |||
| AK215365.1 | - | 1, 2 | 189 | |||
| BC029033.1 | - | 11, 12 | 1675 | |||
| BC029217.1 | - | 7, 8 | 1085 | |||
| BC033452.1 | - | 14, 15 | 2132 | 3156622 | ||

Slc30a5
Genomic Map