siRNA Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022885.2 | NP_075023.2 | 15 | 2315 | ||
| GenBank mRNA | AF233321.1 | - | 8 | 1334 | ||
| AF461761.1 | - | 15 | 2214 | |||
| AK051183.1 | - | 15 | 2315 | |||
| AK088004.1 | - | 14 | 2059 | |||
| AK090176.1 | - | 15 | 2224 | |||
| AK166006.1 | - | 15 | 2125 | |||
| BC029033.1 | - | 12 | 1757 | |||
| BC029217.1 | - | 8 | 1167 | |||
| BC033452.1 | - | 15 | 2214 | 3156622 | ||

Slc30a5
Genomic Map