siRNA Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022885.2 | NP_075023.2 | 4 | 556 | ||
| GenBank mRNA | AF461761.1 | - | 4 | 455 | ||
| AK051183.1 | - | 4 | 556 | |||
| AK088004.1 | - | 4 | 471 | |||
| AK090176.1 | - | 4 | 465 | |||
| AK166006.1 | - | 4 | 366 | |||
| BC033452.1 | - | 4 | 455 | 3156622 | ||

Slc30a5
Genomic Map