siRNA Details
Gene Symbol: Pycr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133705.2 | NP_598466.1 | 7 | 1108 | ||
GenBank mRNA | AK007535.1 | - | 7 | 1089 | ||
AK150756.1 | - | 5 | 2228 | |||
AK152302.1 | - | 5 | 2228 | |||
AK165618.1 | - | 7 | 1083 | |||
AK167622.1 | - | 7 | 1064 | |||
AK179626.1 | - | 2 | 149 | |||
AK186434.1 | - | 2 | 147 | |||
BC006882.1 | - | 7 | 1084 | 2647612 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
