siRNA Details
| Gene Symbol: Chac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026929.4 | NP_081205.1 | 3 | 698 | ||
| GenBank mRNA | AK007378.1 | - | 3 | 651 | ||
| AK032218.1 | - | 3 | 684 | |||
| BC025169.1 | - | 3 | 639 | 4483043 | ||
| BI872476.1 | - | 3 | 575 | |||

Chac1
Genomic Map