siRNA Details
| Gene Symbol: 1810010H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163473.1 | NP_001156945.1 | 3 | 808 | ||
| GenBank mRNA | AK007422.1 | - | 3 | 911 | ||
| BB826454.1 | - | 1 | 137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1810010H24Rik| Gene Symbol: 1810010H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163473.1 | NP_001156945.1 | 3 | 808 | ||
| GenBank mRNA | AK007422.1 | - | 3 | 911 | ||
| BB826454.1 | - | 1 | 137 | |||