siRNA Details
| Gene Symbol: Gmppa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133708.3 | NP_598469.1 | 9 | 893 | ||
| GenBank mRNA | AK007463.1 | - | 9 | 880 | ||
| AK144311.1 | - | 9 | 1158 | |||
| AK150473.1 | - | 10 | 1029 | |||
| AK188808.1 | - | 1 | 29 | |||
| BC008116.1 | - | 9 | 852 | 3498021 | ||

Gmppa
Genomic Map