siRNA Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026934.3 | NP_081210.2 | 9, 10 | 1166 | ||
| GenBank mRNA | AK007452.1 | - | 4, 5 | 397 | ||
| AK139511.1 | - | 9, 10 | 1262 | |||
| AK152374.1 | - | 9, 10 | 1257 | |||
| AK167696.1 | - | 9, 10 | 1259 | |||
| BC028787.1 | - | 4, 5 | 605 | 1246186 | ||
| BC058229.1 | - | 9, 10 | 1093 | 6313954 | ||

Zc3h15
Genomic Map