siRNA Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026935.4 | NP_081211.3 | 6 | 759 | ||
| GenBank mRNA | AK007384.1 | - | 6 | 619 | ||
| AK144336.1 | - | 6 | 699 | |||
| AY005469.1 | - | 6 | 759 | |||
| BC022665.1 | - | 6 | 752 | 4223678 | ||

Sult1c2
Genomic Map