siRNA Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199187.2 | NP_001186116.1 | 4 | 368 | ||
| NM_026937.3 | NP_081213.1 | 4 | 381 | |||
| GenBank mRNA | AK007519.1 | - | 4 | 365 | ||
| AK053811.1 | - | 4 | 382 | |||
| AK139944.1 | - | 4 | 368 | |||
| AK144614.1 | - | 4 | 342 | |||
| AK171958.1 | - | 4 | 342 | |||
| BB844213.1 | - | 4 | 362 | |||
| BC030905.1 | - | 4 | 347 | 4503445 | ||

Ascc1
Genomic Map