siRNA Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199187.2 | NP_001186116.1 | 7 | 783 | ||
| NM_026937.3 | NP_081213.1 | 7 | 796 | |||
| GenBank mRNA | AK007519.1 | - | 7 | 780 | ||
| AK139944.1 | - | 7 | 783 | |||
| AK171958.1 | - | 7 | 757 | |||
| BC030905.1 | - | 7 | 762 | 4503445 | ||

Ascc1
Genomic Map