siRNA Details
| Gene Symbol: Ubr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160319.1 | NP_001153791.1 | 29 | 4098 | ||
| GenBank mRNA | AB093242.2 | - | 2 | 205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubr4| Gene Symbol: Ubr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160319.1 | NP_001153791.1 | 29 | 4098 | ||
| GenBank mRNA | AB093242.2 | - | 2 | 205 | ||