siRNA Details
| Gene Symbol: Adh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026945.1 | NP_081221.1 | 6, 7 | 891 | ||
| GenBank mRNA | AK007397.1 | - | 6, 7 | 887 | ||
| AK028114.1 | - | 6, 7 | 893 | |||
| BC130016.1 | - | 6, 7 | 860 | 40140468 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adh6a| Gene Symbol: Adh6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026945.1 | NP_081221.1 | 6, 7 | 891 | ||
| GenBank mRNA | AK007397.1 | - | 6, 7 | 887 | ||
| AK028114.1 | - | 6, 7 | 893 | |||
| BC130016.1 | - | 6, 7 | 860 | 40140468 | ||