siRNA Details
| Gene Symbol: Eci3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026947.4 | NP_081223.1 | 9 | 904 | ||
| GenBank mRNA | AK007578.1 | - | 9 | 905 | ||
| AK131590.1 | - | 9 | 905 | |||
| BC014724.1 | - | 9 | 878 | 4219360 | ||
| FJ687155.1 | - | 8 | 827 | |||

Eci3
Genomic Map