siRNA Details
| Gene Symbol: 1810022K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099674.1 | NP_001093144.1 | 2 | 144 | ||
| GenBank mRNA | AK007585.1 | - | 2 | 145 | ||
| BC045157.1 | - | 2 | 131 | |||
| BC055766.1 | - | 2 | 132 | |||
| BC083104.1 | - | 2 | 143 | |||
| BC100487.1 | - | 2 | 140 | |||
| BC100488.1 | - | 2 | 128 | |||
| BC115807.1 | - | 2 | 302 | 40106016 | ||

1810022K09Rik
Genomic Map