siRNA Details
| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026956.2 | NP_081232.2 | 7 | 601 | ||
| GenBank mRNA | AK007638.1 | - | 7 | 658 | ||
| BC038121.1 | - | 8 | 632 | |||
| BC046292.1 | - | 5 | 642 | |||
| BC052782.1 | - | 7 | 592 | |||
| BC099497.1 | - | 7 | 596 | |||

Cd209f
Genomic Map