siRNA Details
| Gene Symbol: Kbtbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164574.1 | NP_001158046.1 | 3 | 1283 | ||
| NM_026962.3 | NP_081238.2 | 4 | 1315 | |||
| GenBank mRNA | AK034334.1 | - | 4 | 1223 | ||
| AK078744.1 | - | 3 | 1195 | |||
| AK083497.1 | - | 4 | 1297 | |||
| BC027392.1 | - | 4 | 1219 | 4954712 | ||

Kbtbd3
Genomic Map