siRNA Details
| Gene Symbol: Snx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080557.2 | NP_542124.1 | 6 | 638 | ||
| GenBank mRNA | AK007706.1 | - | 6 | 680 | ||
| AK030657.1 | - | 6 | 651 | |||
| AK031368.1 | - | 650 | ||||
| AK135356.1 | - | 6 | 645 | |||
| BC006043.1 | - | 6 | 628 | |||
| BC016599.1 | - | 6 | 638 | 2644991 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001127550, BC166791.

Snx4
Genomic Map