siRNA Details
| Gene Symbol: Snx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080557.2 | NP_542124.1 | 1, 2 | 153 | ||
| GenBank mRNA | AK007706.1 | - | 1, 2 | 195 | ||
| AK030657.1 | - | 1, 2 | 166 | |||
| AK031368.1 | - | 166 | ||||
| AK135356.1 | - | 1, 2 | 160 | |||
| BC006043.1 | - | 1, 2 | 143 | |||
| BC016599.1 | - | 1, 2 | 153 | 2644991 | ||

Snx4
Genomic Map