siRNA Details
| Gene Symbol: Lzic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026963.5 | NP_081239.1 | 2 | 721 | ||
| GenBank mRNA | AK007657.1 | - | 2 | 210 | ||
| AK146824.1 | - | 2 | 788 | |||
| AK210899.1 | - | 1 | 51 | |||
| BC023174.1 | - | 2 | 721 | 4952948 | ||
| DQ058008.1 | - | 1 | 46 | |||

Lzic
Genomic Map