siRNA Details
| Gene Symbol: Lzic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026963.5 | NP_081239.1 | 3 | 794 | ||
| GenBank mRNA | AK007657.1 | - | 3 | 283 | ||
| AK146824.1 | - | 3 | 861 | |||
| BC023174.1 | - | 3 | 794 | 4952948 | ||
| DQ058008.1 | - | 2 | 119 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK161071.1 | - | 376 | Chr. - - [-] on Build | ||

Lzic
Genomic Map