siRNA Details
| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028839.4 | NP_083115.3 | 4 | 493 | ||
| GenBank mRNA | AK007740.1 | - | 4 | 501 | ||
| AK009341.1 | - | 4 | 472 | |||
| AK076341.1 | - | 4 | 472 | |||
| AK149088.1 | - | 4 | 466 | |||
| AK155257.1 | - | 4 | 458 | |||
| BC024583.1 | - | 2 | 181 | 5065836 | ||

Tmem110
Genomic Map