siRNA Details
| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076341.1 | - | 7 | 790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem110| Gene Symbol: Tmem110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK076341.1 | - | 7 | 790 | ||