siRNA Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027727.2 | NP_082003.1 | 2 | 306 | ||
| GenBank mRNA | AK016957.1 | - | 2 | 235 | ||
| AK049909.1 | - | 2 | 271 | |||
| AK080149.1 | - | 2 | 283 | |||
| AK081242.1 | - | 2 | 298 | |||
| AK149628.1 | - | 2 | 280 | |||
| AK149879.1 | - | 2 | 281 | |||
| AK149891.1 | - | 2 | 280 | |||
| AK154794.1 | - | 2 | 295 | |||
| AK217117.1 | - | 1 | 50 | |||
| BC038276.1 | - | 2 | 308 | 4458792 | ||
| BC092283.1 | - | 2 | 292 | |||

Dym
Genomic Map