siRNA Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027727.2 | NP_082003.1 | 6, 7 | 760 | ||
| GenBank mRNA | AK016957.1 | - | 6, 7 | 689 | ||
| AK049909.1 | - | 6, 7 | 725 | |||
| AK080149.1 | - | 6, 7 | 737 | |||
| AK081242.1 | - | 6, 7 | 752 | |||
| AK149628.1 | - | 6, 7 | 734 | |||
| AK149879.1 | - | 6, 7 | 735 | |||
| AK149891.1 | - | 6, 7 | 734 | |||
| AK154794.1 | - | 6, 7 | 749 | |||
| BC018220.1 | - | 2, 3 | 120 | 3985017 | ||
| BC038276.1 | - | 6, 7 | 762 | 4458792 | ||
| BC092283.1 | - | 6, 7 | 746 | |||

Dym
Genomic Map