siRNA Details
| Gene Symbol: Tmem121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153776.2 | NP_722471.2 | 2 | 887 | ||
| GenBank mRNA | AF488729.1 | - | 2 | 862 | ||
| AK010454.2 | - | 2 | 865 | |||
| BC023493.1 | - | 1 | 269 | |||
| BC043474.1 | - | 1 | 844 | 5364389 | ||

Tmem121
Genomic Map