siRNA Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093752.1 | NP_001087221.1 | 12 | 1299 | ||
| NM_001093753.1 | NP_001087222.1 | 11 | 1281 | |||
| GenBank mRNA | AK081511.1 | - | 9 | 1106 | ||
| AK137854.1 | - | 4 | 373 | |||
| AK141191.1 | - | 5 | 411 | |||
| AK146719.1 | - | 11 | 1253 | |||
| AK183815.1 | - | 3 | 177 | |||
| BC055763.1 | - | 13 | 1186 | |||
| BC138145.1 | - | 9 | 1034 | 8861164 | ||
| BC145162.1 | - | 8 | 893 | 9053713 | ||

Srsf11
Genomic Map