siRNA Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148927.2 | NP_683729.2 | 17 | 1923 | ||
| GenBank mRNA | AK010410.1 | - | 1 | 23 | ||
| AK088969.1 | - | 1 | 23 | |||
| BC024961.1 | - | 16 | 1968 | 4945500 | ||
| BC030731.1 | - | 11 | 1127 | |||
| BE652135.1 | - | 5 | 430 | |||

Plekha4
Genomic Map