siRNA Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148927.2 | NP_683729.2 | 6 | 515 | ||
| GenBank mRNA | BC021387.1 | - | 6 | 526 | 5055226 | |
| BC024961.1 | - | 4 | 474 | 4945500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekha4| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148927.2 | NP_683729.2 | 6 | 515 | ||
| GenBank mRNA | BC021387.1 | - | 6 | 526 | 5055226 | |
| BC024961.1 | - | 4 | 474 | 4945500 | ||