siRNA Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148927.2 | NP_683729.2 | 20 | 2611 | ||
| GenBank mRNA | AK010410.1 | - | 3 | 598 | ||
| AK088969.1 | - | 3 | 598 | |||
| BC024961.1 | - | 18 | 2606 | 4945500 | ||
| BC030731.1 | - | 13 | 1765 | |||
| BC034903.1 | - | 2 | 1408 | |||

Plekha4
Genomic Map