siRNA Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040396.2 | NP_001035486.2 | 4 | 491 | ||
| GenBank mRNA | AK013022.1 | - | 4 | 455 | ||
| AK032838.1 | - | 4 | 468 | |||
| AK047958.1 | - | 4 | 469 | |||
| AK146778.1 | - | 4 | 379 | |||
| BC019970.1 | - | 4 | 439 | 4458171 | ||
| BC038867.1 | - | 4 | 367 | 1547418 | ||
| BC043068.1 | - | 377 | ||||
| BC132453.1 | - | 4 | 457 | 40130730 | ||
| BC138424.1 | - | 4 | 457 | 8861444 | ||

Selt
Genomic Map