siRNA Details
| Gene Symbol: Selt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040396.2 | NP_001035486.2 | 5 | 620 | ||
| GenBank mRNA | AK013022.1 | - | 5 | 584 | ||
| AK032838.1 | - | 5 | 597 | |||
| AK047958.1 | - | 5 | 598 | |||
| AK146778.1 | - | 5 | 508 | |||
| BC019970.1 | - | 5 | 568 | 4458171 | ||
| BC038867.1 | - | 5 | 496 | 1547418 | ||
| BC043068.1 | - | 506 | ||||
| BC132453.1 | - | 5 | 586 | 40130730 | ||
| BC138424.1 | - | 5 | 586 | 8861444 | ||

Selt
Genomic Map